Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcb9
Entrez Gene ID
56325
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 9
Gene Aliases
TAPL, mKIAA1520
Chromosome Location
Chr.5: 124061857 - 124095966 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019875.2 NP_063928.2 4 - 5 1070 133
GenBank mRNA AB045382.2 - 3 - 4 890 133
AB093298.1 - 4 - 5 1052 133
AF216495.1 - 4 - 5 1063 133
AK044140.1 - 4 - 5 1048 133
BC053014.1 - 4 - 5 1023 133

View Full Product Details