Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mphosph9
Entrez Gene ID
269702
Gene Name
M-phase phosphoprotein 9
Gene Aliases
4930548D04Rik, 9630025B04Rik, AW413446, AW547060, B930097C17Rik, C87456, MPP-9, MPP9
Chromosome Location
Chr.5: 124250959 - 124330759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081323.2 NP_001074792.1 18 - 19 3167 91
NM_001277867.1 NP_001264796.1 19 - 20 3257 91
NR_102690.1 19 - 20 3213 91
GenBank mRNA BC076624.1 - 18 - 19 3030 91
BC138423.1 - 15 - 16 2569 91
BC145681.1 - 15 - 16 2569 91

View Full Product Details