Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx5
Entrez Gene ID
16873
Gene Name
LIM homeobox protein 5
Gene Aliases
Lim2
Chromosome Location
Chr.5: 120431886 - 120441457 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008499.5 NP_032525.1 4 - 5 1232 79
GenBank mRNA AK033927.1 - 4 - 5 1219 79
AK048778.1 - 4 - 5 1221 79
AK140208.1 - 4 - 5 1233 79
BC057585.1 - 4 - 5 1120 79
U61155.1 - 4 - 5 1348 79

View Full Product Details