The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019805.4 | NP_062779.3 | 10 - 11 | 1644 | 64 |
GenBank mRNA | AF076607.2 | - | 10 - 11 | 1608 | 64 |
AK030775.1 | - | 10 - 11 | 1609 | 64 | |
AK150764.1 | - | 10 - 11 | 1608 | 64 | |
AK150891.1 | - | 10 - 11 | 1608 | 64 | |
AK153114.1 | - | 10 - 11 | 1608 | 64 | |
AK159523.1 | - | 10 - 11 | 1576 | 64 | |
AK164202.1 | - | 10 - 11 | 1613 | 64 | |
BC006635.1 | - | 10 - 11 | 1597 | 64 |