Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atxn2
Entrez Gene ID
20239
Gene Name
ataxin 2
Gene Aliases
9630045M23Rik, ATX2, AW544490, Sca2
Chromosome Location
Chr.5: 121711609 - 121814950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009125.2 NP_033151.2 24 - 25 3862 76
GenBank mRNA AK135879.1 - 13 - 14 1969 76
AK135976.1 - 19 - 20 2819 76

View Full Product Details