The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145542.3 | NP_663517.2 | 16 - 17 | 1859 | 92 |
GenBank mRNA | AB092504.1 | - | 16 - 17 | 1588 | 92 |
AK078823.1 | - | 9 - 10 | 1061 | 92 | |
BC018218.1 | - | 16 - 17 | 1688 | 92 |