Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdap1
Entrez Gene ID
231887
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
Chromosome Location
Chr.5: 145128770 - 145140089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033313.3 NP_001028485.1 3 - 4 326 102
GenBank mRNA AK147170.1 - 3 - 4 327 102
AK161518.1 - 2 - 3 455 102
AK169233.1 - 3 - 4 260 103
BC007162.1 - 2 - 3 226 102
BC055927.1 - 3 - 4 240 102
BC106168.1 - 2 - 3 214 102
BC139204.1 - 3 - 4 279 102
BC139205.1 - 3 - 4 279 102

View Full Product Details