Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Khdrbs3
Entrez Gene ID
13992
Gene Name
KH domain containing, RNA binding, signal transduction associated 3
Gene Aliases
Etle, SLM-2, Salp, Slm2, T-STAR
Chromosome Location
Chr.15: 68928405 - 69101163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010158.2 NP_034288.2 7 - 8 1220 82
GenBank mRNA AF079763.1 - 7 - 8 1228 82
AF099092.1 - 7 - 8 1231 82
AK014353.1 - 7 - 8 1206 82
AK132792.1 - 7 - 8 922 82
AK160564.1 - 4 - 5 567 82
BC031507.1 - 7 - 8 966 82
BC057577.1 - 7 - 8 1204 82

View Full Product Details