Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Khdrbs3
- Entrez Gene ID
-
13992
- Gene Name
- KH domain containing, RNA binding, signal transduction associated 3
- Gene Aliases
-
Etle, SLM-2, Salp, Slm2, T-STAR
- Chromosome Location
-
Chr.15: 68928405 - 69101163 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014353.1 |
- |
9 - 10 |
1414 |
70 |
View Full Product Details