The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285434.1 | NP_001272363.1 | 2 - 3 | 220 | 61 |
GenBank mRNA | BC013059.1 | - | 2 - 3 | 168 | 61 |