The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008437.1 | NP_032463.1 | 3 - 4 | 507 | 63 |
GenBank mRNA | AB038144.1 | - | 3 - 4 | 520 | 63 |
AK002520.1 | - | 3 - 4 | 354 | 63 | |
AK007861.1 | - | 3 - 4 | 353 | 63 | |
AK152663.1 | - | 3 - 4 | 345 | 63 | |
AK170013.1 | - | 3 - 4 | 351 | 63 | |
BC014813.1 | - | 3 - 4 | 340 | 63 | |
D88899.1 | - | 3 - 4 | 507 | 63 |