The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039562.1 | NP_001034651.1 | 4 - 5 | 688 | 73 |
GenBank mRNA | AK020869.1 | - | 4 - 5 | 672 | 73 |
AK075868.1 | - | 4 - 5 | 562 | 73 | |
BC039642.1 | - | 4 - 5 | 686 | 73 | |
BC092377.1 | - | 4 - 5 | 688 | 73 |