Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpm6a
Entrez Gene ID
234267
Gene Name
glycoprotein m6a
Gene Aliases
Gpm6, M6A
Chromosome Location
Chr.8: 54779434 - 55060878 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253754.1 NP_001240683.1 5 - 6 792 83
NM_001253756.1 NP_001240685.1 4 - 5 970 83
NM_153581.6 NP_705809.1 5 - 6 1163 83
GenBank mRNA AK053538.1 - 5 - 6 717 83
AK081877.1 - 5 - 6 1049 83
AK147245.1 - 5 - 6 950 83
AK147280.1 - 5 - 6 950 83
BC023461.1 - 4 - 5 590 83
BC024759.1 - 5 - 6 690 83
BC029683.1 - 5 - 6 732 83
BC029711.1 - 5 - 6 686 83
BC033357.1 - 5 - 6 732 83
BC033394.1 - 5 - 6 690 83
BC043130.1 - 5 - 6 978 83
CD354193.1 - 4 - 5 579 83
CX564488.1 - 4 - 5 584 83
S65735.1 - 5 - 6 689 83

View Full Product Details