Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
60527
Gene Name
fatty acid desaturase 3
Gene Aliases
AI464531
Chromosome Location
Chr.19: 10040855 - 10059671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021890.3 NP_068690.3 4 - 5 816 70
GenBank mRNA AA981959.1 - 1 - 2 105 70
AK029318.1 - 4 - 5 675 70
AK080414.1 - 4 - 5 668 70
AK090042.1 - 4 - 5 798 70
AK154534.1 - 4 - 5 693 70
AK172509.1 - 4 - 5 795 70

View Full Product Details