The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008886.2 | NP_032912.2 | 13 - 14 | 2326 | 145 |
GenBank mRNA | AK083049.1 | - | 13 - 14 | 2283 | 145 |
AK146365.1 | - | 13 - 14 | 2339 | 145 | |
BC022179.1 | - | 12 - 13 | 2126 | 145 | |
BC058189.1 | - | 13 - 14 | 2332 | 145 | |
BC066110.1 | - | 13 - 14 | 2326 | 145 | |
BC083502.1 | - | 13 - 14 | 2286 | 145 | |
BC086637.1 | - | 13 - 14 | 2302 | 145 | |
BC110684.1 | - | 12 - 13 | 2954 | 145 | |
BC139774.1 | - | 13 - 14 | 1760 | 145 | |
BC141287.1 | - | 13 - 14 | 2302 | 145 |