Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Utp23
- Entrez Gene ID
-
78581
- Gene Name
- UTP23, small subunit (SSU) processome component, homolog (yeast)
- Gene Aliases
-
1700010I21Rik, AI662478, BB238373, D530033C11Rik
- Chromosome Location
-
Chr.15: 51877441 - 51884622 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC027540.1 |
- |
1 - 2 |
207 |
84 |
View Full Product Details