The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164607.1 | NP_001158079.1 | 3 - 4 | 545 | 76 |
NM_001164608.1 | NP_001158080.1 | 2 - 3 | 452 | 76 | |
NM_026859.3 | NP_081135.3 | 2 - 3 | 499 | 76 | |
GenBank mRNA | AK004406.1 | - | 2 - 3 | 172 | 76 |
AK042913.1 | - | 2 - 3 | 393 | 76 | |
AK154047.1 | - | 2 - 3 | 442 | 76 | |
BC016260.1 | - | 2 - 3 | 487 | 76 | |
CK619143.1 | - | 3 - 4 | 490 | 76 |