Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apol7a
Entrez Gene ID
75761
Gene Name
apolipoprotein L 7a
Gene Aliases
9130022K13Rik, Apol3
Chromosome Location
Chr.15: 77388217 - 77399110 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164640.1 NP_001158112.1 2 - 3 262 84
NM_029419.2 NP_083695.2 3 - 4 362 84
GenBank mRNA AK018646.1 - 3 - 4 364 84
AK085956.1 - 2 - 3 239 84
BC139137.1 - 2 - 3 141 84
BC139138.1 - 2 - 3 141 84

View Full Product Details