The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010953.2 | NP_035083.1 | 7 - 8 | 678 | 85 |
GenBank mRNA | AF091846.1 | - | 5 - 6 | 532 | 85 |
AF093591.1 | - | 7 - 8 | 788 | 85 | |
AK012981.1 | - | 7 - 8 | 680 | 85 |