Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam20c
Entrez Gene ID
80752
Gene Name
family with sequence similarity 20, member C
Gene Aliases
C76981, DMP-4, DMP4, GEF-CK, mKIAA4081
Chromosome Location
Chr.5: 138754306 - 138810072 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030565.6 NP_085042.2 9 - 10 2040 68
GenBank mRNA AK149780.1 - 10 - 11 2120 68
AK220286.1 - 9 - 10 1556 68
AY778962.1 - 9 - 10 2021 68
BC004044.1 - 6 - 7 649 68
BC025814.1 - 9 - 10 1991 68
BC025826.1 - 9 - 10 2021 68
CT010363.1 - 5 - 6 495 68

View Full Product Details