The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044747.2 | NP_001038212.1 | 1 - 2 | 138 | 60 |
NM_001163797.1 | NP_001157269.1 | 1 - 2 | 138 | 60 | |
NM_013844.2 | NP_038872.1 | 1 - 2 | 138 | 60 | |
GenBank mRNA | AB024005.1 | - | 1 - 2 | 122 | 60 |
AK013868.1 | - | 1 - 2 | 133 | 60 | |
AK044831.1 | - | 1 - 2 | 108 | 60 | |
AK146092.1 | - | 1 - 2 | 105 | 60 | |
AK153887.1 | - | 1 - 2 | 74 | 60 | |
BC018171.1 | - | 1 - 2 | 123 | 60 | |
BY088951.1 | - | 1 - 2 | 141 | 60 |