Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nudt1
- Entrez Gene ID
-
17766
- Gene Name
- nudix (nucleoside diphosphate linked moiety X)-type motif 1
- Gene Aliases
-
Mth1
- Chromosome Location
-
Chr.5: 140321627 - 140338137 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC021940.1 |
- |
1 - 2 |
60 |
60 |
View Full Product Details