Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyth4
- Entrez Gene ID
-
72318
- Gene Name
- cytohesin 4
- Gene Aliases
-
2510004M07Rik, 5830469K17Rik, AI467541, Pscd4, mFLJ00017
- Chromosome Location
-
Chr.15: 78597047 - 78622019 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC018505.1 |
- |
2 - 3 |
144 |
67 |
View Full Product Details