Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc15a4
Entrez Gene ID
100561
Gene Name
solute carrier family 15, member 4
Gene Aliases
AA987064, AW742963, C130069N12Rik, PHT1, PTR4
Chromosome Location
Chr.5: 127595664 - 127632907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133895.1 NP_598656.1 3 - 4 1031 110
GenBank mRNA AK048529.1 - 3 - 4 998 110
AK154238.1 - 3 - 4 1000 110
AK177058.1 - 1 - 2 64 110
AK188034.1 - 1 - 2 64 110
AK188139.1 - 1 - 2 64 110
BC016233.1 - 3 - 4 1031 110

View Full Product Details