Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbp8
Entrez Gene ID
76074
Gene Name
guanylate-binding protein 8
Gene Aliases
5830443L24Rik, mGBP8
Chromosome Location
Chr.5: 105012513 - 105053609 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029509.4 NP_083785.3 - 1485 101
GenBank mRNA BC031475.1 - - 2402 101
BC080665.1 - 5 - 6 925 101
DQ295175.1 - 8 - 9 1400 101

View Full Product Details