Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgfbr3
Entrez Gene ID
21814
Gene Name
transforming growth factor, beta receptor III
Gene Aliases
1110036H20Rik, AU015626, AW215636, TBRIII
Chromosome Location
Chr.5: 107106570 - 107289595 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011578.3 NP_035708.2 5 - 6 1003 103
GenBank mRNA AF039601.1 - 5 - 6 803 103
AK133234.1 - 3 - 4 511 103
AK166082.1 - 5 - 6 1004 103
BC070428.1 - 5 - 6 928 103

View Full Product Details