The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110140.3 | NP_001103610.1 | 10 - 11 | 1832 | 72 |
NM_009722.3 | NP_033852.1 | 10 - 11 | 1832 | 72 | |
NR_027838.1 | 10 - 11 | 1832 | 72 | ||
GenBank mRNA | AF039893.1 | - | 6 - 7 | 979 | 72 |
AJ131821.1 | - | 10 - 11 | 1502 | 72 | |
AJ223584.1 | - | 10 - 11 | 1502 | 72 | |
AK220538.1 | - | 10 - 11 | 1781 | 72 | |
BC054531.1 | - | 10 - 11 | 1491 | 72 | |
BC054748.1 | - | 10 - 11 | 1491 | 72 |