Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp2a2
Entrez Gene ID
11938
Gene Name
ATPase, Ca++ transporting, cardiac muscle, slow twitch 2
Gene Aliases
9530097L16Rik, D5Wsu150e, SERCA2, SERCA2B, Serca2a, mKIAA4195
Chromosome Location
Chr.5: 122453513 - 122502225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110140.3 NP_001103610.1 11 - 12 1956 124
NM_009722.3 NP_033852.1 11 - 12 1956 124
NR_027838.1 11 - 12 1956 124
GenBank mRNA AF039893.1 - 7 - 8 1103 124
AJ131821.1 - 11 - 12 1626 124
AJ223584.1 - 11 - 12 1626 124
AK210339.1 - 1 - 2 80 124
AK220538.1 - 11 - 12 1905 124
BC054531.1 - 11 - 12 1615 124
BC054748.1 - 11 - 12 1615 124

View Full Product Details