The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039478.1 | NP_001034567.1 | 1 - 2 | 149 | 124 |
GenBank mRNA | DN174290.1 | - | 1 - 2 | 149 | 124 |