Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Agfg2
Entrez Gene ID
231801
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
A630095P14Rik, Hrbl, RAB-R, RABR
Chromosome Location
Chr.5: 137650463 - 137684806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK155221.1 - 2 - 3 415 88
AK171048.1 - 2 - 3 460 88
BC003330.1 - 2 - 3 427 88
BC046788.1 - 2 - 3 351 88

View Full Product Details