Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxn4
Entrez Gene ID
116810
Gene Name
forkhead box N4
Gene Aliases
-
Chromosome Location
Chr.5: 114254164 - 114274598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148935.2 NP_683737.2 9 - 10 1377 71
GenBank mRNA AF323488.1 - 9 - 10 1373 71
AK081735.1 - 7 - 8 1082 71
AY039039.1 - 9 - 10 1423 71
BC092242.1 - 9 - 10 1366 71
BC130222.1 - 9 - 10 1378 71

View Full Product Details