Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pus1
Entrez Gene ID
56361
Gene Name
pseudouridine synthase 1
Gene Aliases
A730013B20Rik, MPUS1, mPus1p
Chromosome Location
Chr.5: 110773667 - 110781056 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025562.2 NP_001020733.1 1 - 2 59 60

View Full Product Details