Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rac1
Entrez Gene ID
19353
Gene Name
RAS-related C3 botulinum substrate 1
Gene Aliases
AL023026, D5Ertd559e
Chromosome Location
Chr.5: 143505481 - 143528116 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK166383.1 - 3 - 4 429 67

View Full Product Details