Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlc1
Entrez Gene ID
50768
Gene Name
deleted in liver cancer 1
Gene Aliases
A730069N07Rik, Arhgap7, HP, STARD12, dlc-1
Chromosome Location
Chr.8: 36567739 - 36953165 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194940.2 NP_001181869.1 1 - 2 321 65
GenBank mRNA AK043205.1 - 1 - 2 303 65
AK045326.1 - 1 - 2 316 65
AK053142.1 - 1 - 2 236 65

View Full Product Details