Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brat1
Entrez Gene ID
231841
Gene Name
BRCA1-associated ATM activator 1
Gene Aliases
AA881470, Baat1
Chromosome Location
Chr.5: 140704786 - 140719379 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276287.1 NP_001263216.1 12 - 13 1781 54
NM_181066.3 NP_851411.1 12 - 13 1683 54
NR_074085.1 11 - 12 1591 54
GenBank mRNA AF226663.1 - 2 - 3 230 54
AK049321.1 - 11 - 12 1580 54
AK084987.1 - 12 - 13 1672 54
AK085091.1 - 12 - 13 1672 54
AK154448.1 - 12 - 13 1654 54
AK170269.1 - 12 - 13 1677 54
BC068301.1 - 12 - 13 1747 54

View Full Product Details