Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tchp
Entrez Gene ID
77832
Gene Name
trichoplein, keratin filament binding
Gene Aliases
A930031F18Rik, AV279199
Chromosome Location
Chr.5: 114707765 - 114722327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029992.2 NP_084268.2 5 - 6 708 65
GenBank mRNA AK083027.1 - 5 - 6 624 65
AK159947.1 - 5 - 6 669 65
BC043048.1 - 5 - 6 613 65

View Full Product Details