Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt4
Entrez Gene ID
75387
Gene Name
sirtuin 4
Gene Aliases
4930596O17Rik
Chromosome Location
Chr.5: 115478006 - 115490050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133760.1 NP_598521.1 1 - 2 150 66

View Full Product Details