Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc64
Entrez Gene ID
75665
Gene Name
coiled-coil domain containing 64
Gene Aliases
2210403N09Rik, BICDR-1
Chromosome Location
Chr.5: 115648174 - 115731643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080808.1 NP_001074277.1 4 - 5 919 82
GenBank mRNA AK036757.1 - 1 - 2 379 82
BC024804.1 - 4 - 5 912 82
BC126948.1 - 4 - 5 919 82
GU973876.1 - 4 - 5 919 82

View Full Product Details