The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080808.1 | NP_001074277.1 | 8 - 9 | 1569 | 100 |
GenBank mRNA | AK170295.1 | - | 2 - 3 | 178 | 100 |
AK204022.1 | - | 1 - 2 | 131 | 100 | |
BC024804.1 | - | 8 - 9 | 1562 | 100 | |
BC049161.1 | - | 2 - 3 | 307 | 100 | |
BC126948.1 | - | 8 - 9 | 1569 | 100 | |
GU973876.1 | - | 8 - 9 | 1569 | 100 |