Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Amz1
- Entrez Gene ID
-
231842
- Gene Name
- archaelysin family metallopeptidase 1
- Gene Aliases
-
5330426I05, 6530401C20Rik
- Chromosome Location
-
Chr.5: 140724127 - 140753312 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173405.2 |
NP_775581.1 |
2 - 3 |
140 |
108 |
View Full Product Details