The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008494.3 | NP_032520.1 | 7 - 8 | 1146 | 62 |
GenBank mRNA | AF015768.1 | - | 7 - 8 | 1175 | 62 |
AI116033.1 | - | 2 - 3 | 157 | 62 | |
AK004642.1 | - | 7 - 8 | 62 | ||
AK153283.1 | - | 7 - 8 | 1147 | 62 | |
BC115680.1 | - | 6 - 7 | 812 | 62 | |
BC138600.1 | - | 7 - 8 | 1167 | 62 | |
BC138601.1 | - | 7 - 8 | 1167 | 62 | |
U94351.1 | - | 7 - 8 | 1111 | 62 |