Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cabp5
Entrez Gene ID
29865
Gene Name
calcium binding protein 5
Gene Aliases
-
Chromosome Location
Chr.7: 13383295 - 13408880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB079605.1 - 1 - 2 60 80

View Full Product Details