Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgms1
Entrez Gene ID
208449
Gene Name
sphingomyelin synthase 1
Gene Aliases
9530058O11Rik, AI841905, C80702, Mob, Sms1, Sor1, Tmem23
Chromosome Location
Chr.19: 32122727 - 32389705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168525.1 NP_001161997.1 6 - 7 1800 85
NM_001168526.1 NP_001161998.1 2 - 3 1024 85
NM_144792.4 NP_659041.3 6 - 7 1368 85
GenBank mRNA AK079254.1 - 2 - 3 969 85
AK133967.1 - 2 - 3 992 85
AK146782.1 - 6 - 7 1368 85
AK150272.1 - 2 - 3 1001 85
AK151170.1 - 6 - 7 1353 85
AK152312.1 - 2 - 3 1002 85
AK153406.1 - 6 - 7 1353 85
AK160539.1 - 2 - 3 1002 85
AY509044.1 - 6 - 7 1368 85
BC019443.1 - 2 - 3 991 85
BC076618.1 - 6 - 7 1800 85
BC085298.1 - 3 - 4 962 85

View Full Product Details