Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dkkl1
Entrez Gene ID
50722
Gene Name
dickkopf-like 1
Gene Aliases
AI414495, SGY, SGY-1, SGY1, Soggy, mSgy
Chromosome Location
Chr.7: 45207525 - 45212602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015789.3 NP_056604.2 2 - 3 301 94
GenBank mRNA AF177399.1 - 2 - 3 206 94
AF274312.1 - 2 - 3 260 94
AK131694.1 - 2 - 3 246 94
BC002215.1 - 2 - 3 279 94
CA466481.1 - 2 - 3 304 94

View Full Product Details