Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oas2
Entrez Gene ID
246728
Gene Name
2'-5' oligoadenylate synthetase 2
Gene Aliases
Oasl11
Chromosome Location
Chr.5: 120730333 - 120749884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145227.3 NP_660262.2 9 - 10 2085 62
GenBank mRNA AB067535.1 - 9 - 10 2090 62
AF418010.1 - 9 - 10 2025 62
AK049218.1 - 1 - 2 599 62
BC043667.1 - 3 - 4 542 62
BC118538.1 - 9 - 10 2026 62

View Full Product Details