Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasgef1b
Entrez Gene ID
320292
Gene Name
RasGEF domain family, member 1B
Gene Aliases
4732452O09Rik, GPIgamma4, Gpig4
Chromosome Location
Chr.5: 99217420 - 99252927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145839.2 NP_665838.1 1 - 2 209 115
NM_181318.4 NP_851835.1 1 - 2 209 115
GenBank mRNA AK028756.1 - 1 - 2 101 115
AK090085.1 - 1 - 2 101 115
AK149812.1 - 1 - 2 128 115
AY129963.1 - 1 - 2 136 115
BC050858.1 - 1 - 2 142 115
BC052726.1 - 1 - 2 100 115
BY130572.1 - 1 - 2 210 115

View Full Product Details