Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coq2
Entrez Gene ID
71883
Gene Name
coenzyme Q2 homolog, prenyltransferase (yeast)
Gene Aliases
2310002F18Rik, PHB:PPT
Chromosome Location
Chr.5: 100654723 - 100674285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027978.2 NP_082254.2 4 - 5 637 88
GenBank mRNA AK009092.1 - 4 - 5 639 88
BC080773.1 - 4 - 5 632 88

View Full Product Details