The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081107.1 | NP_001074576.1 | 2 - 3 | 1017 | 144 |
GenBank mRNA | AK052427.1 | - | 2 - 3 | 977 | 144 |
BC082601.1 | - | 2 - 3 | 981 | 144 | |
BC109169.2 | - | 2 - 3 | 885 | 144 | |
BC109170.2 | - | 2 - 3 | 890 | 144 |