Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pprc1
- Entrez Gene ID
-
226169
- Gene Name
- peroxisome proliferative activated receptor, gamma, coactivator-related 1
- Gene Aliases
-
A930018M19, BC013720, PRC, mKIAA0595
- Chromosome Location
-
Chr.19: 46056496 - 46072909 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK044522.1 |
- |
4 - 5 |
580 |
80 |
View Full Product Details