The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007383.3 | NP_031409.2 | 9 - 10 | 1162 | 50 |
GenBank mRNA | AK155361.1 | - | 9 - 10 | 1141 | 50 |
AK169428.1 | - | 9 - 10 | 1099 | 50 | |
BC016259.1 | - | 9 - 10 | 1129 | 50 | |
BC055063.1 | - | 8 - 9 | 1315 | 50 |